Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Aspartic aciddiethyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Asp(OEt)-OEt.HClL-Aspartic acid diethyl ester hydrochlorideH-Asp(OEt)-OEt·HClDiethyl L-Aspartate hydrochloride

H63974-22 H63974-14 H63974-06
AAH63974-06EA 31.39 USD InStock
AAH63974-06 AAH63974-14 AAH63974-22
L-Aspartic aciddiethyl ester hydrochloride 98%
L-Aspartic aciddiethyl ester hydrochloride


Formula: C₈H₁₆ClNO₄
MW: 225.669 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038910
CAS Number: 16115-68-7
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty