Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Hyp-OMe 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl esterN-Cbz-4-trans-hydroxy-L-proline methyll esterN-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyll esterZ-Hyp-OMe(2S,4R)-1-Benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylateN-Carbobenzoxy-4-trans-hydroxy-L-proline methyll ester

H63993-06 H63993-MD H63993-03
AAH63993-03EA 75.81 USD InStock
AAH63993-03 AAH63993-MD AAH63993-06
Z-Hyp-OMe 98%
Z-Hyp-OMe


Formula: C₁₄H₁₇NO₅
MW: 279.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055851
CAS Number: 64187-48-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty