Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-fluoro-5-nitrobenzene 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-1-fluoro-4-nitrobenzene3-Bromo-4-fluoronitrobenzene

H64045-22 H64045-14
AAH64045-14EA 53.89 USD InStock
AAH64045-14 AAH64045-22
1-Bromo-2-fluoro-5-nitrobenzene 95%
1-Bromo-2-fluoro-5-nitrobenzene


Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055433
CAS Number: 701-45-1
EINECS: 211-855-6
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty