Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4'-(trifluoromethoxy)acetophenone 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(Trifluoromethoxy)phenacyl bromide2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone

H64105-06 H64105-03 H64105-MD
AAH64105-MDEA 25.69 USD InStock
AAH64105-MD AAH64105-03 AAH64105-06
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone


Formula: C₉H₆BrF₃O₂
MW: 283.04 g/mol
Melting Pt: 48…55 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052333
CAS Number: 103962-10-3
UN: 3261
ADR: "8",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty