Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-Bromo-5-fluorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Bromo-2-chloro-5-fluorobenzaldehyde2-Chloro-4-bromo-5-fluorobenzaldehyde

H64238-MD H64238-06 H64238-03
AAH64238-06EA 265.99 USD InStock
AAH64238-06 AAH64238-MD AAH64238-03
2-Chloro-4-Bromo-5-fluorobenzaldehyde 98%
2-Chloro-4-Bromo-5-fluorobenzaldehyde


Formula: C₇H₃BrClFO
MW: 237.452 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185877
CAS Number: 1214386-29-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty