Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyano-2-hydroxypyridine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Cyano-2-pyridone2-Oxo-1,2-dihydropyridine-3-carbonitrile

H64254-22 H64254-06 H64254-14
AAH64254-06EA 36.09 USD InStock
AAH64254-06 AAH64254-14 AAH64254-22
3-Cyano-2-hydroxypyridine 98%
3-Cyano-2-hydroxypyridine


Formula: C6H4N2O
MW: 120.11 g/mol
Melting Pt: 224…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00661865
CAS Number: 20577-27-9
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty