Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-tert-Butylglycine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(tert-Butylamino)acetic acid hydrochloride

H64270-06 H64270-14 H64270-03
AAH64270-14EA 318.99 USD InStock
AAH64270-14 AAH64270-03 AAH64270-06
N-tert-Butylglycine hydrochloride 97%
N-tert-Butylglycine hydrochloride


Formula: C₆H₁₄ClNO₂
MW: 167.633 g/mol
Melting Pt: 223…224 °C
Storage Temperature: Ambient
MDL Number: MFCD03788441
CAS Number: 6939-23-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty