Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-amino-3-iminopropanoate hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethyl 2-amidinoacetate hydrochloride

H64288-03 H64288-14 H64288-06
AAH64288-06EA 105.99 USD InStock
AAH64288-06 AAH64288-03 AAH64288-14
Ethyl-3-amino-3-iminopropanoate hydrochloride 95%
Ethyl-3-amino-3-iminopropanoate hydrochloride


Formula: C5H11ClN2O2
MW: 166.60 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD06797615
CAS Number: 57508-48-2
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty