Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine

H64329-MD H64329-03 H64329-06
AAH64329-03EA 88.59 USD InStock
AAH64329-03 AAH64329-06 AAH64329-MD
2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%
2-chloro-5-iodo-3-(trifluoromethyl)pyridine


Formula: C₆H₂ClF₃IN
MW: 307.439 g/mol
Melting Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD12024302
CAS Number: 887707-25-7
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty