Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-chlorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H64330-06 H64330-14 H64330-22
AAH64330-14EA 83.89 USD InStock
AAH64330-14 AAH64330-22 AAH64330-06
2-Bromo-5-chlorobenzaldehyde 97%
2-Bromo-5-chlorobenzaldehyde


Formula: C₇H₄BrClO
Melting Pt: 64…68 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00462870
CAS Number: 174265-12-4
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty