Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(Trimethylsilylmethyl)benzylamine 95%

Supplier: Thermo Scientific Chemicals
Warning

H64450-06 H64450-22 H64450-14
AAH64450-14EA 136.04 USD InStock
AAH64450-14 AAH64450-22 AAH64450-06
N-(Trimethylsilylmethyl)benzylamine 95%
N-(Trimethylsilylmethyl)benzylamine


Formula: C11H19NSi
MW: 193.36 g/mol
Boiling Pt: 89…90 °C/5 mm
Density: 0.88
Flash Pt: 96 °C(205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012192
CAS Number: 53215-95-5
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty