Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Aminocyclobutanecarboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Amino-1-cyclobutanecarboxylic acid

H64459-06 H64459-03
AAH64459-03EA 44.49 USD InStock
AAH64459-03 AAH64459-06
1-Aminocyclobutanecarboxylic acid 95%
1-Aminocyclobutanecarboxylic acid


Formula: C₅H₉NO₂
MW: 115.132 g/mol
Melting Pt: 261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00661068
CAS Number: 22264-50-2
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty