Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Formylthiophene-2-carbonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Cyanothiophene-2-carboxaldehyde

H64462-MD H64462-06 H64462-03
AAH64462-03EA 112.99 USD InStock
AAH64462-03 AAH64462-MD AAH64462-06
5-Formylthiophene-2-carbonitrile 95%
5-Formylthiophene-2-carbonitrile


Formula: C₆H₃NOS
MW: 137.156 g/mol
Melting Pt: 90…95 °C
Storage Temperature: Ambient
MDL Number: MFCD09037804
CAS Number: 21512-16-3
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty