Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-nitropyrimidin-4-amine 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Amino-2-chloro-5-nitropyrimidine

H64478-06 H64478-MD H64478-03
AAH64478-MDEA 30.6 USD InStock
AAH64478-MD AAH64478-06 AAH64478-03
2-Chloro-5-nitropyrimidin-4-amine 97%
2-Chloro-5-nitropyrimidin-4-amine


Formula: C₄H₃ClN₄O₂
MW: 174.544 g/mol
Melting Pt: 221…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00127771
CAS Number: 1920-66-7
EINECS: 217-648-7
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty