Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxyphenethylamine 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(3-Methoxyphenyl)ethylamine

H64494-06 H64494-22 H64494-14
AAH64494-22EA 399.73 USD InStock
AAH64494-22 AAH64494-14 AAH64494-06
3-Methoxyphenethylamine 97+%
3-Methoxyphenethylamine


Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 118…118 °C/6 mm
Density: 1.038
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008187
CAS Number: 2039-67-0
EINECS: 218-017-9
UN: 2735
ADR: "8",III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty