Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-2-iodobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H64643-06 H64643-14
AAH64643-06EA 48.79 USD InStock
AAH64643-06 AAH64643-14
5-Bromo-2-iodobenzoic acid 97%
5-Bromo-2-iodobenzoic acid


Formula: C₇H₄BrIO₂
Melting Pt: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00079717
CAS Number: 21740-00-1
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty