Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-2-hydroxy-4-picoline 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Bromo-2-hydroxy-4-methylpyridine

H64674-06 H64674-14
AAH64674-06EA 52.19 USD InStock
AAH64674-06 AAH64674-14
5-Bromo-2-hydroxy-4-picoline 97%
5-Bromo-2-hydroxy-4-picoline


Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD03427664
CAS Number: 164513-38-6
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty