Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorophenethylamine 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(3-Fluorophenyl)ethylamine

H64762-14 H64762-06 H64762-22
AAH64762-06EA 38.02 USD InStock
AAH64762-06 AAH64762-22 AAH64762-14
3-Fluorophenethylamine 97+%
3-Fluorophenethylamine


Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 87 °C/15 mm
Density: 1.066
Flash Pt: 83 °C(181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075376
CAS Number: 404-70-6
UN: 2735
ADR: "8",III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty