Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-9,9-dimethylfluorene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 9,9-Dimethyl-9H-fluoren-2-amine

H64771-MD H64771-06 H64771-03
AAH64771-03EA 72.39 USD InStock
AAH64771-03 AAH64771-06 AAH64771-MD
2-Amino-9,9-dimethylfluorene 98%
2-Amino-9,9-dimethylfluorene


Formula: C₁₅H₁₅N
MW: 209.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953790
CAS Number: 108714-73-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty