Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Oxocyclobutanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H64774-06 H64774-14
AAH64774-14EA 242.99 USD InStock
AAH64774-14 AAH64774-06
3-Oxocyclobutanecarboxylic acid 98%
3-Oxocyclobutanecarboxylic acid


Formula: C5H6O3
MW: 114.10 g/mol
Melting Pt: 69…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00100900
CAS Number: 23761-23-1
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty