Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-5-nitrobenzaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

H64787-03 H64787-06 H64787-MD
AAH64787-06EA 487.99 USD InStock
AAH64787-06 AAH64787-03 AAH64787-MD
3-Bromo-5-nitrobenzaldehyde 97+%
3-Bromo-5-nitrobenzaldehyde


Formula: C₇H₄BrNO₃
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00100097
CAS Number: 355134-13-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty