Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(N,N-Diphenylamino)benzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Diphenylaminobenzaldehyde

H64887-06 H64887-03 H64887-14
AAH64887-03EA 29.79 USD InStock
AAH64887-03 AAH64887-14 AAH64887-06
4-(N,N-Diphenylamino)benzaldehyde 98%
4-(N,N-Diphenylamino)benzaldehyde


Formula: C₁₉H₁₅NO
MW: 273.33 g/mol
Melting Pt: 129…133 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00145131
CAS Number: 4181-05-9
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty