Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Bromophenyl)propan-2-ol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(4-Bromophenyl)-2-propanol

H64919-03 H64919-MD
AAH64919-MDEA 62.19 USD InStock
AAH64919-MD AAH64919-03
2-(4-Bromophenyl)propan-2-ol 98%
2-(4-Bromophenyl)propan-2-ol


Formula: C₉H₁₁BrO
Boiling Pt: 75…85° /0.1 mm
Melting Pt: 40…45 °C
Density: 1.356
Storage Temperature: Ambient
MDL Number: MFCD11870096
CAS Number: 2077-19-2
EINECS: 816-112-5
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty