Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-pyrimidineboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloropyrimidine-5-boronic acid

H64949-03 H64949-MD H64949-06
AAH64949-MDEA 25.23 USD InStock
AAH64949-MD AAH64949-03 AAH64949-06
2-Chloro-5-pyrimidineboronic acid 96%
2-Chloro-5-pyrimidineboronic acid


Formula: C₄H₄BClN₂O₂
MW: 158.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08063113
CAS Number: 1003845-06-4
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty