Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Hyp(tBu)-OH 95%

Supplier: Thermo Scientific Chemicals
Synonyms: trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-prolineZ-Hyp(Tbu)-OH

H65039-14 H65039-03 H65039-06
AAH65039-14EA 873.98 USD InStock
AAH65039-14 AAH65039-03 AAH65039-06
Z-Hyp(tBu)-OH 95%
Z-Hyp(tBu)-OH


Formula: C₁₇H₂₃NO₅
MW: 321.373 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153336
CAS Number: 85201-91-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty