Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Gln-Gly-OH 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-2-(5-Amino-2-(((benzyloxy)carbonyl)amino)-5-oxopentanamido)acetic acidN-Benzyloxycarbonyl-L-glutaminylglycine

H65088-03 H65088-MD H65088-06
AAH65088-03EA 176.99 USD InStock
AAH65088-03 AAH65088-MD AAH65088-06
Z-Gln-Gly-OH 98%
Z-Gln-Gly-OH


Formula: C₁₅H₁₉N₃O₆
MW: 337.332 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00055926
CAS Number: 6610-42-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty