Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Histidine methyl ester dihydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-D-His-OMe.2HCl

H65096-06 H65096-03 H65096-14
AAH65096-06EA 102.99 USD InStock
AAH65096-06 AAH65096-03 AAH65096-14
D-Histidine methyl ester dihydrochloride 95%
D-Histidine methyl ester dihydrochloride


Formula: C₇H₁₁N₃O₂
MW: 242.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066137
CAS Number: 4467-54-3
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty