Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Methylamino)benzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H65148-06 H65148-14
AAH65148-06EA 177.99 USD InStock
AAH65148-06 AAH65148-14
4-(Methylamino)benzonitrile 97%
4-(Methylamino)benzonitrile


Formula: C₈H₈N₂
MW: 132.16 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00016381
CAS Number: 4714-62-9
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty