Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Serinamide hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Ser-NH₂·HCl

H65169-22 H65169-14 H65169-06
AAH65169-22EA 1237.83 USD InStock
AAH65169-22 AAH65169-14 AAH65169-06
L-Serinamide hydrochloride 95%
L-Serinamide hydrochloride


Formula: C₃H₉ClN₂O₂
MW: 140.567 g/mol
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00067560
CAS Number: 65414-74-6
EINECS: 265-753-1
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty