Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Ala-Ala-NH₂ 97%

Supplier: Thermo Scientific Chemicals
Synonyms: N-(Benzyloxycarbonyl)alanyl-L-alaninamide

H65212-06 H65212-03
AAH65212-03EA 56.38 USD InStock
AAH65212-03 AAH65212-06
Z-Ala-Ala-NH₂ 97%
Z-Ala-Ala-NH₂


Formula: C₁₄H₁₉N₃O₄
Storage Temperature: Ambient
MDL Number: MFCD00238398
CAS Number: 50444-54-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty