Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl-2-thienylmethylphosphonate 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Diethyl (thiophen-2-ylmethyl)phosphonate

H65232-MD H65232-03 H65232-06
AAH65232-06EA 336.99 USD InStock
AAH65232-06 AAH65232-MD AAH65232-03
Diethyl-2-thienylmethylphosphonate 95%
Diethyl-2-thienylmethylphosphonate


Formula: C₉H₁₅O₃PS
MW: 234.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128457
CAS Number: 2026-42-8
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty