Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloropicolinamide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Chloropyridine-2-carboxamide

H65265-14 H65265-22 H65265-06
AAH65265-14EA 107.99 USD InStock
AAH65265-14 AAH65265-22 AAH65265-06
4-Chloropicolinamide 97%
4-Chloropicolinamide


Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD01085339
CAS Number: 99586-65-9
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty