Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Alaninamide hydrochloride 96%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-Aminopropionamide Hydrochloride

H65365-22 H65365-06 H65365-14
AAH65365-06EA 60.09 USD InStock
AAH65365-06 AAH65365-22 AAH65365-14
β-Alaninamide hydrochloride 96%
beta-Alaninamide hydrochloride


Formula: C₃H₈N₂O
MW: 124.57 g/mol
Melting Pt: 149…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00060747
CAS Number: 64017-81-8
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty