Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-β-Homoglutamic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Synonyms: H-β-HomoGlu-OH·HCl

H65370-MD H65370-03
AAH65370-MDEA 137.64 USD InStock
AAH65370-MD AAH65370-03
L-β-Homoglutamic acid hydrochloride 97%
L-beta-Homoglutamic acid hydrochloride


Formula: C₆H₁₂ClNO₄
MW: 197.615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862872
CAS Number: 61884-74-0
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty