Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Benzyloxycarbonyl-N-methyl-D-valine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Z-D-N-Me-Val-OH

H65531-06 H65531-MD H65531-03
AAH65531-06EA 503.99 USD InStock
AAH65531-06 AAH65531-03 AAH65531-MD
N-Benzyloxycarbonyl-N-methyl-D-valine 95%
N-Benzyloxycarbonyl-N-methyl-D-valine


Formula: C₁₄H₁₉NO₄
MW: 265.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973293
CAS Number: 53978-73-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty