Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Leu-OSu 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Z-Leu-OsuN-Benzyloxycarbonyl-L-leucine N-succinimidyl ester

H65546-22 H65546-06 H65546-14
AAH65546-06EA 50.29 USD InStock
AAH65546-06 AAH65546-14 AAH65546-22
Z-Leu-OSu 95%
Z-Leu-OSu


Formula: C₁₈H₂₂N₂O₆
MW: 362.382 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00042759
CAS Number: 3397-35-1
EINECS: 222-256-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty