Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Cyclopropylglycine 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Cyclopropyl-L-glycine(S)-2-Amino-2-cyclopropylacetic acid

H65755-MD H65755-03 H65755-06
AAH65755-MDEA 58.89 USD InStock
AAH65755-MD AAH65755-06 AAH65755-03
L-Cyclopropylglycine 97%
L-Cyclopropylglycine


Formula: C₅H₉NO₂
MW: 115.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659116
CAS Number: 49606-99-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty