Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Asn-OtBu·HCl 95%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Asn-OtBu·HCl

H65792-06 H65792-MD H65792-03
AAH65792-03EA 90.29 USD InStock
AAH65792-03 AAH65792-MD AAH65792-06
H-Asn-OtBu·HCl 95%
H-Asn-OtBu·HCl


Formula: C₈H₁₇ClN₂O₃
MW: 224.685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039079
CAS Number: 63094-81-5
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty