Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Propargylglycine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-D-Pra-OH

H65798-06 H65798-03
AAH65798-06EA 341.99 USD InStock
AAH65798-06 AAH65798-03
D-Propargylglycine 97%
D-Propargylglycine


Formula: C₅H₇NO₂
MW: 113.116 g/mol
Melting Pt: 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00237407
CAS Number: 23235-03-2
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty