Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-D-alanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-N-Me-D-Ala-OH

H65840-06 H65840-14
AAH65840-14EA 395.99 USD InStock
AAH65840-14 AAH65840-06
N-Methyl-D-alanine 98%
N-Methyl-D-alanine


Formula: C₄H₉NO₂
MW: 103.121 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063135
CAS Number: 29475-64-7
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty