Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Benzyl-D-glutamate 95%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Glutamic Acid 1-Benzyl EsterH-D-Glu-OBzl

H65903-06 H65903-03 H65903-14
AAH65903-03EA 22.53 USD InStock
AAH65903-03 AAH65903-14 AAH65903-06
1-Benzyl-D-glutamate 95%
1-Benzyl-D-glutamate


Formula: C₁₂H₁₅NO₄
MW: 237.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237065
CAS Number: 79338-14-0
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty