Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-((Ethoxythioxomethyl)thio)-2-methylpropanoic acid

H66036-MD H66036-06 H66036-03
AAH66036-03EA 65.6 USD InStock
AAH66036-03 AAH66036-06 AAH66036-MD
2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid 95%
2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid


Formula: C₇H₁₂O₃S₂
MW: 208.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19689494
CAS Number: 133944-74-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty