Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3-bromo-6-methylpyridine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-3-bromo-6-picoline 3-Bromo-6-methyl-2-pyridylamine 3-Bromo-6-methylpyridin-2-amine

H66162-06 H66162-03
AAH66162-06EA 183.99 USD InStock
AAH66162-06 AAH66162-03
2-Amino-3-bromo-6-methylpyridine 95%
2-Amino-3-bromo-6-methylpyridine


Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 78…83 °C
Storage Temperature: Ambient
MDL Number: MFCD03095215
CAS Number: 126325-46-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty