Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydroxymethyloxetane 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,4-Epoxybutanol2-Oxetanemethanol

H66218-03 H66218-MD
AAH66218-MDEA 104.99 USD InStock
AAH66218-MD AAH66218-03
2-Hydroxymethyloxetane 96%
2-Hydroxymethyloxetane


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 98 °C/15 mm
Density: 1.080
Storage Temperature: Freezer
MDL Number: MFCD00142933
CAS Number: 61266-70-4
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty