Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2,6-difluorophenylacetic acid 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(4-Bromo-2,6-difluorophenyl)acetic acid

H66231-03 H66231-MD
AAH66231-MDEA 71.35 USD InStock
AAH66231-MD AAH66231-03
4-Bromo-2,6-difluorophenylacetic acid 96%
4-Bromo-2,6-difluorophenylacetic acid


Formula: C₈H₅BrF₂O₂
MW: 251.027 g/mol
Density: 1.77
Storage Temperature: Ambient
MDL Number: MFCD03094466
CAS Number: 537033-54-8
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty