Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-N-fmoc-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Phe(3-Cl)-OH

H66237-06 H66237-03
AAH66237-03EA 58.79 USD InStock
AAH66237-03 AAH66237-06
3-Chloro-N-fmoc-L-phenylalanine 95%
3-Chloro-N-fmoc-L-phenylalanine


Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672548
CAS Number: 198560-44-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty