Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ɑ-Cyclohexyl-DL-mandelic acid 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: α-Cyclohexylmandelic acid2-Cyclohexyl-2-hydroxy-2-phenylacetic acid

H66238-06 H66238-14
AAH66238-14EA 284.99 USD InStock
AAH66238-14 AAH66238-06
ɑ-Cyclohexyl-DL-mandelic acid 97%
alpha-Cyclohexyl-DL-mandelic acid


Formula: C₁₄H₁₈O₃
MW: 234.30 g/mol
Boiling Pt: 135 °C/3 mm
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00194543
CAS Number: 4335-77-7
EINECS: 224-380-4
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty