Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methoxy-1H-indole-2-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Methoxyindole-2-carboxylic acid

H66239-06 H66239-03
AAH66239-03EA 67.19 USD InStock
AAH66239-03 AAH66239-06
6-Methoxy-1H-indole-2-carboxylic acid 95%
6-Methoxy-1H-indole-2-carboxylic acid


Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 198…203 °C
Storage Temperature: Ambient
MDL Number: MFCD01548823
CAS Number: 16732-73-3
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty