Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(Fmoc-amino)valeric acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-5-Ava-OH

H66264-06 H66264-14 H66264-03
AAH66264-14EA 636.99 USD InStock
AAH66264-14 AAH66264-03 AAH66264-06
5-(Fmoc-amino)valeric acid 98%
5-(Fmoc-amino)valeric acid


Formula: C₂₀H₂₁NO₄
MW: 339.391 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00235889
CAS Number: 123622-48-0
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty