Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Fmoc-L-valinol 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Valinol

H66323-06 H66323-03
AAH66323-03EA 51.19 USD InStock
AAH66323-03 AAH66323-06
N-Fmoc-L-valinol 98%
N-Fmoc-L-valinol


Formula: C₂₀H₂₃NO₃
MW: 325.408 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00235961
CAS Number: 160885-98-3
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty